missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5242506

 View more versions of this product

Catalog No. AAH5242506


Explore available promotions

Description

Description

(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(R)-3-Amino-1-benzylpiperidine
C12H18N2
5 g
Slightly soluble in water.
C1CC(CN(C1)CC2=CC=CC=C2)N
190.29
190.29
168466-84-0
MFCD03093374
r-3-amino-1-benzylpiperidine, r-1-benzylpiperidin-3-amine, r-3-amino-1-benzyl-piperidine, 3r-1-benzylpiperidin-3-amine, r-1-benzyl-3-aminopiperidine, 3-piperidinamine, 1-phenylmethyl-, 3r, s-3-amino-n-benzylpiperidine, pubchem11493, r-n-benzyl-3-aminopiperidine, r-1-benzyl-piperidin-3-ylamine
HARWNWOLWMTQCC-GFCCVEGCSA-N
(3R)-1-benzylpiperidin-3-amine
854130
97%

Chemical Identifiers

168466-84-0
190.29
HARWNWOLWMTQCC-GFCCVEGCSA-N
854130
C1CC(CN(C1)CC2=CC=CC=C2)N
C12H18N2
MFCD03093374
r-3-amino-1-benzylpiperidine, r-1-benzylpiperidin-3-amine, r-3-amino-1-benzyl-piperidine, 3r-1-benzylpiperidin-3-amine, r-1-benzyl-3-aminopiperidine, 3-piperidinamine, 1-phenylmethyl-, 3r, s-3-amino-n-benzylpiperidine, pubchem11493, r-n-benzyl-3-aminopiperidine, r-1-benzyl-piperidin-3-ylamine
(3R)-1-benzylpiperidin-3-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only