Learn More
(R)-3-Amino-3-phenylpropionic acid, 95%
CAS: 83649-48-3 | C9H12ClNO2 | 201.65 g/mol
$111.04 - $1271.01
Chemical Identifiers
| CAS | 83649-48-3 |
|---|---|
| Molecular Formula | C9H12ClNO2 |
| Molecular Weight (g/mol) | 201.65 |
| MDL Number | MFCD01076237 |
| InChI Key | ABEBCTCOPRULFS-DDWIOCJRSA-N |
| Synonym | r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt |
| PubChem CID | 2734697 |
| IUPAC Name | (3R)-3-amino-3-phenylpropanoic acid;hydrochloride |
| SMILES | C1=CC=C(C=C1)C(CC(=O)O)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52034MD
|
Thermo Scientific Chemicals
H52034MD |
250 mg |
Each for $111.04
|
|
|||||
|
AAH5203403
|
Thermo Scientific Chemicals
H5203403 |
1 g |
Each for $326.58
|
|
|||||
|
AAH5203406
|
Thermo Scientific Chemicals
H5203406 |
5 g |
Each for $1,271.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83649-48-3 | |
| 201.65 | |
| ABEBCTCOPRULFS-DDWIOCJRSA-N | |
| 2734697 | |
| C1=CC=C(C=C1)C(CC(=O)O)N.Cl |
| C9H12ClNO2 | |
| MFCD01076237 | |
| r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt | |
| (3R)-3-amino-3-phenylpropanoic acid;hydrochloride |
Specifications
| 83649-48-3 | |
| MFCD01076237 | |
| ABEBCTCOPRULFS-DDWIOCJRSA-N | |
| (3R)-3-amino-3-phenylpropanoic acid;hydrochloride | |
| 201.65 | |
| 165.19 | |
| 95% |
| C9H12ClNO2 | |
| r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt | |
| C1=CC=C(C=C1)C(CC(=O)O)N.Cl | |
| 250 mg | |
| 2734697 | |
| Air Sensitive | |
| (R)-3-Amino-3-phenylpropionic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only