missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-3-Butyn-2-ol, 99%, Thermo Scientific Chemicals

$98.62 - $924.04

Chemical Identifiers

CAS 42969-65-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00211237
InChI Key GKPOMITUDGXOSB-SCSAIBSYSA-N
Synonym r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
PubChem CID 638102
IUPAC Name (2R)-but-3-yn-2-ol
SMILES CC(C#C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL18880MD
View Documents
Thermo Scientific Chemicals
L18880MD
250 mg
Each for $98.62
 
AAL1888003
View Documents
Thermo Scientific Chemicals
L1888003
1 g
Each for $266.62
 
AAL1888006
View Documents
Thermo Scientific Chemicals
L1888006
5 g
Each for $924.04
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

42969-65-3
70.091
GKPOMITUDGXOSB-SCSAIBSYSA-N
638102
CC(C#C)O
C4H6O
MFCD00211237
r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
(2R)-but-3-yn-2-ol

Specifications

42969-65-3
108°C to 109°C
C4H6O
MFCD00211237
UN2929
r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
+47° (Neat)
(2R)-but-3-yn-2-ol
638102
99%
0.891
28°C (82°F)
1.426
250 mg
3587272
GKPOMITUDGXOSB-SCSAIBSYSA-N
CC(C#C)O
70.091
70.09
(R)-(+)-3-Butyn-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H300+H310+H330-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only