Learn More
(R)-(-)-3-Methyl-2-butanol, 98+%
CAS: 1572-93-6 | C5H12O | 88.15 g/mol
$487.22 - $487.22
Chemical Identifiers
| CAS | 1572-93-6 |
|---|---|
| Molecular Formula | C5H12O |
| Molecular Weight (g/mol) | 88.15 |
| MDL Number | MFCD00065949 |
| InChI Key | MXLMTQWGSQIYOW-RXMQYKEDSA-N |
| Synonym | 2r-3-methylbutan-2-ol, r---3-methyl-2-butanol, 2-butanol, 3-methyl-, 2r, r-3-methyl-2-butanol, r-3-methylbutan-2-ol, 2r-3-methyl-2-butanol, r---isopropyl methyl carbinol, unii-2nk7o363q6 component, unii-93ff0f303r component |
| PubChem CID | 638099 |
| IUPAC Name | (2R)-3-methylbutan-2-ol |
| SMILES | CC(C)C(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1100803
|
Thermo Scientific Chemicals
L1100803 |
1 g |
Each for $487.22
|
|
|||||
Description
(R)-(-)-3-Methyl-2-butanol, is used as a pharmaceutical intermediate. It is also used as a solvent and an intermediate in the manufacture of other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(-)-3-Methyl-2-butanol, is used as a pharmaceutical intermediate. It is also used as a solvent and an intermediate in the manufacture of other chemicals.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
| 1572-93-6 | |
| 88.15 | |
| MXLMTQWGSQIYOW-RXMQYKEDSA-N | |
| 638099 | |
| CC(C)C(C)O |
| C5H12O | |
| MFCD00065949 | |
| 2r-3-methylbutan-2-ol, r---3-methyl-2-butanol, 2-butanol, 3-methyl-, 2r, r-3-methyl-2-butanol, r-3-methylbutan-2-ol, 2r-3-methyl-2-butanol, r---isopropyl methyl carbinol, unii-2nk7o363q6 component, unii-93ff0f303r component | |
| (2R)-3-methylbutan-2-ol |
Specifications
| 1572-93-6 | |
| 112°C | |
| C5H12O | |
| MFCD00065949 | |
| UN1105 | |
| 2r-3-methylbutan-2-ol, r---3-methyl-2-butanol, 2-butanol, 3-methyl-, 2r, r-3-methyl-2-butanol, r-3-methylbutan-2-ol, 2r-3-methyl-2-butanol, r---isopropyl methyl carbinol, unii-2nk7o363q6 component, unii-93ff0f303r component | |
| MXLMTQWGSQIYOW-RXMQYKEDSA-N | |
| CC(C)C(C)O | |
| 88.15 | |
| 88.15 | |
| (R)-(-)-3-Methyl-2-butanol |
| 0.81 | |
| 26°C (78°F) | |
| 1.4075 | |
| 1 g | |
| 3535088 | |
| Soluble in alcohol. Insoluble in water. | |
| −5° (Neat) | |
| (2R)-3-methylbutan-2-ol | |
| 638099 | |
| ≥98% |
Safety and Handling
GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only