missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Penten-2-ol, 95%

CAS: 64584-92-5 | C5H10O | 86.13 g/mol

$321.29 - $967.38

Chemical Identifiers

CAS 64584-92-5
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD03701536
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYNA-N
Synonym r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
PubChem CID 6999724
SMILES CC(O)CC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2565903
View Documents
Thermo Scientific Chemicals
B2565903
1 g
Each for $321.29
Only null left
 
AAB2565906
View Documents
Thermo Scientific Chemicals
B2565906
5 g
Each for $967.38
Only null left
 
Description

Description

(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

Solubility
Soluble in water, 4.526 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

64584-92-5
86.13
ZHZCYWWNFQUZOR-UHFFFAOYNA-N
6999724
C5H10O
MFCD03701536
r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
CC(O)CC=C

Specifications

64584-92-5
115°C to 116°C
C5H10O
MFCD03701536
UN1987
Soluble in water,4.526mg/L @ 25°C.
−5.2° (c=1 in Chloroform)
pent-4-en-2-ol
6999724
95%
0.837
30°C (86°F)
1.424
1 g
r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
ZHZCYWWNFQUZOR-UHFFFAOYNA-N
CC(O)CC=C
86.13
86.13
(R)-(-)-4-Penten-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only