missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 264120010

 View more versions of this product

Catalog No. AC264120010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
Specifications

Specifications

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride
98.5
Colorless to Yellow
213.0°C to 214.0°C
Authentic
Glass bottle
1.4680 to 1.4700
1 g
15, 6365
− 132.00
PAORVUMOXXAMPL-VIFPVBQESA-N
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
3080792
98+%
39637-99-5
100.0
1.3400g/mL
89°C
98% min. (GC)
C10H8ClF3O2
MFCD00044400
1.34
−132.00 (20.00°C c=6,CHCl3)
unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
252.62
252.62
Liquid

Chemical Identifiers

39637-99-5
252.62
PAORVUMOXXAMPL-VIFPVBQESA-N
3080792
CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
C10H8ClF3O2
MFCD00044400
unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only