Learn More
(R)-(-)-Carvone, 98%, Thermo Scientific Chemicals
$69.40 - $754.21
Chemical Identifiers
CAS | 6485-40-1 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.22 |
MDL Number | MFCD00001578 |
InChI Key | ULDHMXUKGWMISQ-SECBINFHSA-N |
Synonym | --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone |
PubChem CID | 439570 |
ChEBI | CHEBI:15400 |
SMILES | CC(=C)[C@@H]1CC=C(C)C(=O)C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1390018
|
Thermo Scientific Chemicals
A1390018 |
50 g |
Each for $69.40
|
|
||||||
AAA1390030
|
Thermo Scientific Chemicals
A1390030 |
250 g |
Each for $230.10
|
|
||||||
AAA139000B
|
Thermo Scientific Chemicals
A139000B |
1000 g |
Each for $754.21
|
|
||||||
Description
(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.
Solubility
Miscible with alcohol, ether, chloroform, propylene glycol and mineral oils. Immiscible with water and glycerol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
6485-40-1 | |
150.22 | |
ULDHMXUKGWMISQ-SECBINFHSA-N | |
439570 | |
CC(=C)[C@@H]1CC=C(C)C(=O)C1 |
C10H14O | |
MFCD00001578 | |
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
CHEBI:15400 |
Specifications
6485-40-1 | |
228°C to 230°C | |
98% | |
1.499 | |
50 g | |
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
ULDHMXUKGWMISQ-SECBINFHSA-N | |
(5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one | |
439570 | |
150.22 | |
Liquid |
0.959 | |
93°C (199°F) | |
C10H14O | |
MFCD00001578 | |
2206714 | |
Miscible with alcohol,ether,chloroform,propylene glycol and mineral oils. Immiscible with water and glycerol. | |
CC(=C)[C@@H]1CC=C(C)C(=O)C1 | |
150.22 | |
CHEBI:15400 | |
98% | |
(R)-(-)-Carvone |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
H227-H317
EINECSNumber : 229-352-5
RTECSNumber : OS8650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only