Learn More
(R)-(+)-Limonene, 96%
CAS: 5989-27-5 | C10H16 | 136.24 g/mol
$56.14 - $139.10
Chemical Identifiers
| CAS | 5989-27-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00062991 |
| InChI Key | XMGQYMWWDOXHJM-JTQLQIEISA-N |
| Synonym | d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene |
| PubChem CID | 440917 |
| ChEBI | CHEBI:15382 |
| IUPAC Name | (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene |
| SMILES | CC(=C)[C@@H]1CCC(C)=CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL04733AE
|
Thermo Scientific Chemicals
L04733AE |
100 mL |
N/A
|
|
|||||
|
AAL04733AP
|
Thermo Scientific Chemicals
L04733AP |
500 mL |
N/A
|
|
|||||
Description
Chiral building block. Biodegradable solvent. Limonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLimonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis and as an insect repellant.
Solubility
Miscible with alcohol,chloroform,ether and carbondisulfide.Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Chemical Identifiers
| 5989-27-5 | |
| 136.24 | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| 440917 | |
| (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene |
| C10H16 | |
| MFCD00062991 | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| CHEBI:15382 | |
| CC(=C)[C@@H]1CCC(C)=CC1 |
Specifications
| 5989-27-5 | |
| 0.841 | |
| 48°C (118°F) | |
| C10H16 | |
| MFCD00062991 | |
| UN2052 | |
| 14,5493 | |
| Miscible with alcohol,chloroform,ether and carbondisulfide.Immiscible with water. | |
| +115° (c=10 in Ethanol) | |
| (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene | |
| 440917 | |
| 136.23 | |
| (R)-(+)-Limonene, stabilized |
| -74°C | |
| 177°C to 178°C | |
| Fruit-like | |
| 1.473 | |
| 100 mL | |
| 2204754 | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| CC(=C)[C@@H]1CCC(C)=CC1 | |
| 136.24 | |
| CHEBI:15382 | |
| 96% |
Safety and Handling
GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c
H226-H304-H315-H317-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE
EINECSNumber : 227-813-5
RTECSNumber : GW6360000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only