missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(+)-α-Methylbenzyl isocyanate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(R)-(+)-a-Methylbenzyl isocyanate [(R)-(+)-MBIC] is a derivatizing agent.
$331.70 - $1244.16
Chemical Identifiers
MDL Number | MFCD00063015 |
---|---|
Synonym | (R)-(+)-1-Phenylethyl isocyanate |
Chemical Identifiers
MFCD00063015 | |
(R)-(+)-1-Phenylethyl isocyanate |
Specifications
33375-06-3 | |
55°C to 56°C/2.5 mmHg | |
Glass Bottle | |
MFCD00063015 | |
(R)-(+)-1-Phenylethyl isocyanate | |
≥99.0% (sum of enantiomers, GC); ≥99.0% |
1.045 g/mL at 20°C; 1.046 g/mL (at 20°C) | |
65°C - Closed Cup | |
C6H5 CH(CH3)NCO | |
1 mL | |
147.17 | |
For chiral derivatization |