missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-N-BOC-4-Chlorophenylalanine, 95%, 98% ee, Thermo Scientific™
$140.68 - $140.68
Chemical Identifiers
CAS | 57292-44-1 |
---|---|
Molecular Formula | C14H18ClNO4 |
Molecular Weight (g/mol) | 299.75 |
MDL Number | MFCD00076978 |
InChI Key | BETBOAZCLSJOBQ-LLVKDONJSA-N |
Synonym | boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl |
PubChem CID | 7018791 |
IUPAC Name | (2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC362920010
|
Thermo Scientific Chemicals
362920010 |
1 g |
Each for $140.68
|
|
|||||
Chemical Identifiers
57292-44-1 | |
299.75 | |
BETBOAZCLSJOBQ-LLVKDONJSA-N | |
7018791 | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O |
C14H18ClNO4 | |
MFCD00076978 | |
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl | |
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
57292-44-1 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
MFCD00076978 | |
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O | |
299.75 | |
299.75 | |
Powder |
94.0 | |
110°C | |
97.5% min. (HPLC) | |
94% min. (HPLC) | |
C14H18ClNO4 | |
1 g | |
BETBOAZCLSJOBQ-LLVKDONJSA-N | |
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
7018791 | |
95%,98% ee | |
(R)-N-BOC-4-Chlorophenylalanine, 95%, (98% e.e.) |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only