Epredia™ RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Hold up to 10 slides back-to-back with Epredia™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Supplier:  Epredia™ E94
Catalog No. E94
Explore available promotions
Description
Description
Features
  • Heavy, clear glass construction with internal grooves
  • 1.6 in. diameter circular tapered top
  • Includes drop-on lid
Specifications
Specifications
Yes
Round
Coplin Staining Jar
1.6 in.
Drop-on lid
Glass
10