Learn More
RAM Scientific Safe-T-Fill™ Blood Gas Capillary Tube Caps
End caps for Safe-T-Fill™ blood gas capillaries
Supplier:  RAM Scientific 060402
Catalog No. 14-915-78
Only null left
Explore available promotions
Description
Description
End Caps safely seal SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes, minimizing the risk of blood spillage and allow for easier handling.
  • Tapered end caps are made of PVC and contain no natural rubber latex
  • Ø 6.7mm (width) x 12mm (length)
  • Fits capillary tubes with outer diameters 1.6 - 2.1mm
  • Packaging labels are marked with Lot Number and Expiration Date
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
End Cap
PVC
Safely seals capillary tubes, minimizing the risk of blood spillage and allows for easier handling and processing.
SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes
Tube Caps
Natural
Non-sterile
Product Suggestions
Product Suggestions