RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: EDTA
No scooping needed, allowing for high-quality, tissue-free samples
Supplier: RAM Scientific 076011
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis,
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
- Capillary straw and microtube are coated with anticoagulant Dipotassium (K2) EDTA (Purple cap) to prevent clotting
- Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
- Lot Number, Expiration Date, and fill lines are printed directly on the microtube
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- For whole blood hematology determinations
- Small sample sizes, great for pediatrics and neonates
- Available with self-sealing rubber cap (Not made with natural rubber latex)
- Available in a variety of sizes and draw volumes
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Clear | |
K2 EDTA | |
Non-sterile | |
125 μL | |
Purple | |
Intended for hematology (haematology) determinations that require whole blood samples. |
Hematology/Whole Blood Tube | |
Plastic | |
Standard Pierceable Cap | |
Yes | |
50 Tubes/Bag, 10 Bags/Cs. |