RAM Scientific Safet-T-Fil™ Blood Gas Capillaries
100% plastic—no glass to break
Supplier:  RAM Scientific 060185
 View more versions of this product
Catalog No. 14-915-72
Explore available promotions
Description
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes through the same capillary action as glass without the risk of breakage. Capillary tubes are available untreated or heparinized.
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Replaces glass Natelson, Caraway, or mylar-coated glass blood gas capillary tubes
  • Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
  • Capillary tubes are available untreated or heparinized
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • Optional end caps minimize the risk of blood spillage and allow for easier handling
  • Optional mixing fleas and magnetic rings (mixing components) allow for uniform mixing to prevent clotting
  • Packaging labels are marked with Lot Number and Expiration Date
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
Clear
Balanced Heparin
Non-sterile
Intended for blood gas analysis, tests for multiple analytes, or as a transfer device for PKU and POC testing and coated with Balanced Heparin anticoagulant to prevent clotting.
Blood Gas Tube
Plastic
115 μL
Videos
SDS
Documents
Documents
Accessories