DWK Life Sciences Kimble™ Ramsbottom Coking Bulb
The Ramsbottom Coking Bulb is used in the determination of carbon residue in petroleum products.
Supplier:  DWK Life Sciences 8966500025
Catalog No. 13-756-800
Explore available promotions
Description
Description
  • Ref: ASTM D524
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Ramsbottom Coking Bulb