missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Raney™-Nickel, Activated Catalyst, 50% slurry in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 395921000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Raney™-Nickel | |
7440-02-0,7732-18-5,7429-90-5 | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Plastic bottle |
Activated Catalyst | |
Ni | |
100 g | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
58.69 | |
Slurry |
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Safety and Handling
EINECSNumber : 231-111-4
RUO – Research Use Only