Stoelting™ Rat Adaptor Kit, 18 Degree for Rat Stereotaxic Series 51600 DFS Item
Nose clamp assembly and ear bars are included.
Supplier:  Stoelting™ 51621
Catalog No. 10-000-148
Explore available promotions
Description
Description
  • 30 mm of vertical travel along dovetail slide and 0.1 mm vernier scales
  • 50 mm of Anterior-Posterior travel possible
Specifications
Specifications
Neuroscience
18° ear bars
Rat
Standard ear bars