Kent Scientific Rat Retractor Set for SurgiSuite Multi-Functional Surgical Platforms DFS Item
For use with SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms.
Supplier:  Kent Scientific SURGI5002
Catalog No. 13005181
Explore available promotions
Includes: Two 6cm tall magnetic fixators with four locking jaws, ten 14cm wire retractors, ten retractor tips (two 1mm sharp tip, two 1mm blunt tip, two 2.5mm blunt tip, two 5mm blunt tip, two 7.5mm blunt tip)
Description
Description
  • Designed specifically for Rat
Specifications
Specifications
Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
Retractor Set
SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Two 6cm tall magnetic fixators with four locking jaws, ten 14cm wire retractors, ten retractor tips (two 1mm sharp tip, two 1mm blunt tip, two 2.5mm blunt tip, two 5mm blunt tip, two 7.5mm blunt tip)