Learn More
Eisco™ Raw Phyllite, Metamorphic Rock Specimen - Approx. 3 Inch
Phyllite Specimen for demonstrations of metamorphic properties, such as glossy sheen and dense, foliated minerals.
Supplier:  Eisco™ ESNG0059HAND
Catalog No. S02920
Only null left
Explore available promotions
Description
Description
  • Phyllite is a good example of low-grade metamorphism from an easily recognizable parent rock. The fissile property of slate is retained, but has attained a glossy sheen and soapy feel more indicative of metamorphic material
  • Ideal for identification and classification exercises. Demonstrates characteristic metamorphic properties, such as a glossy sheen and dense, foliated minerals
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Our rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Each specimen sample is packaged in a transparent bag, and is labeled and barcoded for easy identification
  • Each specimen is selected and hand processed by an actual geologist, ensuring that each sample exhibits defining features
  • Rocks and minerals are sourced from a variety of locations around the world and are processed by hand in Victor, NY
Specifications
Specifications
Metamorphic Rock
Product Suggestions
Product Suggestions