Spectrum Brands Rayovac™ Alkaline Maximum™ Industrial Batteries
Ideal for all high-drain, frequent-use devices
Supplier:  Spectrum Brands ALC
 View more versions of this product
Catalog No. 17-926B
Explore available promotions
Description
Description
  • Sealed in steel — guaranteed not to leak
  • Mercury- and cadmium-free
  • Seven-year shelf life
Specifications
Specifications
Alkaline Battery
For commercial applications
50 mm
26.2 mm
C
1.9 in.
1 in.
7500 mAh