Invitrogen™ eBioscience™ 1X RBC Lysis Buffer
Formulated for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood.
Supplier:  Invitrogen™ 00433357
Catalog No. 501129751
Explore available promotions
Description
Description
  • Contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes when used as instructed
  • Nucleated red cells are not effectively lysed with ammonium chloride
Specifications
Specifications
200 mL
Lysis Buffer
eBioscience™
Liquid
200 mL
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.