missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RD Plastics Color Coded Ziplock Biohazard Specimen Bags
Color-coded 6 x 9 in. bags for easy identification. Available in green, red, pink or purple.
Supplier: RD Plastics Q770
Description
Color coded biohazard/specimen transport bag with tear top and paperwork pouch- 2 mil clear reclosable bag with tear-away top for easy opening
- 3-wall construction for durable, reliable protection
- Biohazard symbol printed in black ink on colored block
Specifications
Not Autoclavable | |
Zip Seal | |
Specimen transport | |
LDPE | |
100 | |
9 in. |
1.5 lb. | |
Pink | |
Paperwork pouch | |
1000 | |
22.86 x 15.25cm | |
6 in. |