RD Plastics Three-wall Reclosable Biohazard Specimen Transport Bags
Resealable ziplock closure
Supplier:  RD Plastics RDB22
 View more versions of this product
Catalog No. 23-700-209
Explore available promotions
Description
Description
  • Three-wall construction
  • Pouch has a flap cover to hold paperwork stays in place
  • With orange and black biohazard symbol and storage instructions
Specifications
Specifications
Biohazard Specimen Transport Bag
Used for specimen transport
0.279 mm
9 x 6 in. (22.86 x 15.24cm)
22.86 cm
15.24 cm
Clear
Pouch with flap cover, Biohazard symbol and storage instructions
2 mil
9 in.
6 in.