missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RD Plastics Color Coded Ziplock Biohazard Specimen Bags
Opaque colored specimen transport bags feature a reclosable seal with a tear away top for easy opening. Available in yellow, green, blue or red.
$159.00 - $204.62
Specifications
Product Type | Transport Bag |
---|---|
Autoclavable | Not Autoclavable |
Capacity (English) | 1.5 lb. |
Closure Type | Tear-away Zip-top |
For Use With (Application) | Specimen transport |
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
23-700-231
|
N/A | Yellow |
Case of 1000 Each for $188.07
|
|
||||||
23-700-232
|
RD Plastics
Q865 |
Green |
Case of 1000 Each for $159.00
|
|
||||||
23-700-234
|
N/A | Red |
Case of 1000 Each for $204.62
|
|
||||||
23-700-233
|
N/A | Blue |
Case of 1000 Each for $188.07
|
|
||||||
Description
- LDPE bags in 3 Wall style with 2 mil thickness
- Printed with black "SPECIMEN" and biohazard markings
- Red option also printed with "STAT"
- Color coded for easy specimen handling protocols
- Includes a paperwork pouch
Specifications
Transport Bag | |
1.5 lb. | |
Specimen transport | |
LDPE | |
1000 | |
2 mil | |
9 in. | |
6 in. |
Not Autoclavable | |
Tear-away Zip-top | |
Biohazard print, paperwork pouch | |
0.279 mm | |
100 | |
9 x 6 in. (22.8 x 15.2cm) | |
22.8 cm | |
15.2 cm |