Thermo Scientific™ Reacti-Vap™ Evaporators
Add evaporation to the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module for derivatization or other small scale reactions.
$2342.30 - $5902.72
Specifications
Product Type Evaporator
Temperature (Metric) Ambient +10° to 200°C
View More Specs
Products 2
Catalog Number Mfr. No. Ports For Use With Price Quantity  
Catalog Number Mfr. No. Ports For Use With Price Quantity  
PI18825
Accessories View Documents Promotion Details
Thermo Scientific™
TS18825
9-Ports Single-Block Reacti-Therm Modules
Each for $2,342.30
PI18826
Accessories View Documents Promotion Details
Thermo Scientific™
TS18826
27-Ports Triple-Block Reacti-Therm Modules
Each for $5,902.72
Description
Description
Perform simple, efficient evaporation using Thermo Scientific Reacti-Vap Evaporators. This precision-machined gas manifold performs simultaneous or separate delivery of nonreactive pressurized gas to samples in the Thermo Scientific Reacti-Vap Therm module. The evaporator simply attaches to the Reacti-Therm module. The evaporating head tilts back for easy needle attachment and removal.
PTFE-coated needles are available for applications using strong acids. Each Reacti-Vap Needle has a Luer-Lok™ hub for leak-proof attachment to Reacti-Vap Evaporators. Needles are available in 4- and 6-inch lengths. Reacti-Vap PTFE-Coated Needles, made exclusively for use in Reacti-Vap Evaporators, are blunt-ended, 19-gauge, stainless steel needles that reduce cross-contamination and corrosion when evaporating solvents that contain strong acids.
Ideal applications
  • General sample incubation and evaporation in a variety of tube and vial sizes
  • Silylation, alkylation, and acylation derivatization reactions for GC sample preparation.
Reacti-Therm Sample Derivatization System
The complete derivatization system combines heating, stirring and evaporation in a versatile, modular system that is easily configured to your application. The system comprises (Sold Separately):
  • Reacti-Therm Heating/Stirring Module
  • Reacti-Vap Evaporator Module
  • Reacti-Blocks, Reacti-Vial Magnetic Stirrers, and Reacti-Vial Small Reaction Vials
Specifications
Specifications
Evaporator
Ambient +10° to 200°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.