missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Reading Glass Magnifiers

Three sizes for quick, convenient magnification.

$13.85 - $15.79

Specifications

Material Glass
Product Type Magnifying Glass
For Use With (Application) For quick, convenient magnification
Includes Chrome plated bakelite handle
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W x H) Magnification Weight (English) Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Magnification Weight (English) Weight (Metric) Price Quantity  
S95954
View Documents
Eisco™
PH0510F
7.4 x 6.2 x 3.14 in. (19 x 16 x 8cm) 2.25x 2.2 lb. 1 kg
Each for $15.79
 
S95953
View Documents
Eisco™
PH0510B
6.2 x 6.2 x 2.75 in. (16 x 16 x 7cm) 3x 1.47 lb. 0.67 kg
Each for $13.85
 
Description

Description

  • Chrome-plated metal frames with plastic handles
  • Packed in cardboard box

Specifications

Specifications

Glass
Magnifying Glass
For quick, convenient magnification
Chrome plated bakelite handle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'