missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Reagent Alcohol, 80%, Fisherbrand™, HistoPrep™
Supplier: Fisherbrand HC14001GL
Specifications
Liquid | |
76°C | |
Poly Bottle | |
1 gal. | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 |
Reagent Alcohol, 80% | |
64-17-5,7732-18-5,67-63-0,67-56-1 | |
Yellow | |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 |
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |