missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Reagent Alcohol, 95%, Fisherbrand™, Histoprep™

Supplier: Fisherbrand HC13001GL
Specifications
Liquid | |
-114.1°C | |
64-17-5,7732-18-5,67-56-1,67-63-0 | |
Polyethylene Container | |
Colorless | |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 |
Alcohol, 95% | |
78.5°C | |
0.822g/cm³ | |
40mmHg | |
3.79 L | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 |
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |