missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute, Ricca Chemical

Supplier:  Ricca Chemical Company 659032

 View more versions of this product

Catalog No. 6590-32


Explore available promotions

Description

Description

Specifications

Specifications

Alcohol, Reagent , Anhyd., Absolute
64-17-5,67-63-0,67-56-1
91.0,6.0,5.0
1 L
max. 0.001%
max. 0.5 %
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
ACS
Denatured alcohol
90.0,4.0,4.0
Colorless Liquid
Natural Poly Bottle
C2H6O
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236

Chemical Identifiers

C2H6O
702
ethanol
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
CHEBI:16236
CCO
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS
Documents

Documents

Promotions

Promotions