Learn More
Reagent Alcohol, Reagent Grade, 90.5% Ethanol ±2.0%, LabChem™
Supplier: LabChem LC222004
Specifications
| Reagent Alcohol | |
| 64-17-5,67-63-0,67-56-1 | |
| 90,5.5,4.5 | |
| 790kg/m3 | |
| 4 L | |
| Passes Test | |
| Liquid | |
| C2H6O | |
| UN1987 | |
| Soluble in water | |
| CCO | |
| 46.069 | |
| CHEBI:16236 | |
| 90.5% ±2.0% Ethanol |
| -115°C | |
| Colorless | |
| Carbon monoxide; Carbon dioxide; May release flammable gases | |
| 78°C | |
| 13°C | |
| Poly Bottle | |
| 790kg/m3 | |
| C2H6O | |
| ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol | |
| 702 | |
| 46.07 | |
| Reagent |
Chemical Identifiers
| 64-17-5 | |
| 46.069 | |
| ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
| CHEBI:16236 | |
| CCO |
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| 702 | |
| ethanol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
Causes
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Danger
EINECSNumber : 200-578-6
Recommended Storage : Room Temperature