Promega RealTime-Glo™ MT Cell Viability Assay DFS Item
The RealTime-Glo™ MT Cell Viability Assay is a non-lytic, homogeneous, bioluminescent method to determine in real time the number of viable cells in culture by measuring the reducing potential of cells and thus metabolism (MT).
Supplier:  Promega G9711
 View more versions of this product
Catalog No. PRG9711
Explore available promotions
Description
Description
  • Monitor Cell Viability in Real Time
  • Save time, cell culture and reagent costs
  • Rapidly detect changes in cell viability
Specifications
Specifications
-20°C. Avoid prolonged exposure to light. Avoid multiple freeze-thaw cycles.
100 reactions
Product Suggestions
Product Suggestions