Promega RealTime-Glo™ MT Cell Viability Assay
The RealTime-Glo™ MT Cell Viability Assay is a non-lytic, homogeneous, bioluminescent method to determine in real time the number of viable cells in culture by measuring the reducing potential of cells and thus metabolism (MT).
Supplier: Promega G9713
Description
- Monitor Cell Viability in Real Time
- Save time, cell culture and reagent costs
- Rapidly detect changes in cell viability
Specifications
-20°C. Avoid prolonged exposure to light. Avoid multiple freeze-thaw cycles. | |
1000 reactions |