Fisherbrand™ Recordall™ Chart Paper
Chart Paper for Discontinued Fisher Recordall™ Series 5000 Recorders
 View more versions of this product
Catalog No. 13-939-38
Explore available promotions
Description
Description
  • Roll is 11 in. wide (28cm) and 100 ft. long (30m)
  • Major Division is numbered along the amplitude axis for all units except Cat. No. 13-939-37
† For use with models having electric integrator.
Specifications
Specifications
Continuous Record Chart Paper
For Fisher Recordall™ series 5000 recorders
30.48 m
Major divisions every 1 in.; 6 minor divisions/major
100 ft.
Product Suggestions
Product Suggestions