missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Rectangular Conference Table

Rectangular conference table with a sturdy and stylish base and wire management channels.

Supplier:  Fisherbrand™ FSUP12048

 View more versions of this product

Catalog No. 06001065


Explore available promotions

Description

Description

  • Conference table with 1 in. thick work surface
  • Laminate gray oak color
Specifications

Specifications

120 in.
73.7 cm
121.9 cm
29 in.
48 in.
Plastic Laminate
304.8 cm
Videos
SDS
Documents

Documents

Promotions

Promotions