Fisherbrand™ Rectangular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
Supplier: Fisherbrand™ 24X551602811T
Description
These cover glasses feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in a pack of 1 oz.
- Made of colorless borosilicate glass
- Available in a variety of thicknesses and sizes
Specifications
Coverslip | |
Borosilicate Glass | |
55 x 24 mm | |
1 oz./Pk. |
Rectangle | |
0.13 to 0.17 mm (#1) | |
Microscopy, Histology, Cytology, Hematology, Microbiology |