missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recykleen™ Sharps Collection Trolley
$1328.54 - $1972.59
Specifications
| Color | Gray |
|---|---|
| Sterility | Non-sterile |
| Includes | Liquid-absorbent material |
| For Use With (Application) | Design that reduces the risk of collectors tipping over |
| Material | Recycled Material |
| Catalog Number | Mfr. No. | Capacity (English) | Product Type | Height (Metric) | Width (Metric) | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Product Type | Height (Metric) | Width (Metric) | Price | Quantity | ||
|
22-204-436
|
BD
305092 |
17 to 19 gal. | Standard Trolley | 61.2 cm | 45.7 cm |
Each for $1,328.54
|
|
||
|
22-204-435
|
BD
305091 |
9 gal. | Foot-operated Trolley | 59.6 cm | 41.4 cm |
Each for $1,972.59
|
|
||
Description
These collectors are made from a minimum 70% recycled material and are intended for Resource Conservation and Recovery Act (RCRA) solid hazardous waste. As an added feature, each RCRA collector comes with a liquid-absorbent material preattached to the inside bottom of the collector.
Specifications
| Gray | |
| Liquid-absorbent material | |
| Recycled Material |
| Non-sterile | |
| Design that reduces the risk of collectors tipping over |