MilliporeSigma™ Reflectometer Rqflex™ 20
Read out Reflectoquant™ test strips with the portable RQFlex™ 20 reflectometer.
Supplier:  MilliporeSigma™ 1172460001
Catalog No. M1172460001
Explore available promotions
Description
Description
  • The successor of the RQFlex™ 10 features a redesigned intuitive user interface paired with high accuracy.
  • It allows for fast and easy measurement of a broad range of chemical parameters in the food, water, and environmental applications.
  • Detection Method: Reflectometric
Specifications
Specifications
Reflectoquant™ Test Strips
Fiber Case
Reflectometer
Product Suggestions
Product Suggestions