DR Instruments Reflex Hammer
Reflex Hammer is primary used to test tendon reflexes. A proper tool to perform a neurological examination and testing the integrity of the patellar tendon and other reflexes.
Supplier:  DR Instruments 75
Catalog No. S72118
Explore available promotions
Alert:
California Proposition 65 WARNING: Cancer and Reproductive Harm
Description
Description
  • DR Instruments Reflex Hammer is primary used to test tendon reflexes such as patellar tendon just below the knee, the Achilles tendon in the ankle or other tendon points on the body.
  • It generates stretch reflex response in the muscle, and user assesses the strength and symmetry of the patient's reflex responses.
Specifications
Specifications
Reflex Hammer,Reflex Hammer
Rubber Head, Stainless Steel Handle
Testing patellar and other reflexes