Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rotor Adapters
For 6 x 85mL fixed-angle rotors
Supplier:  Eppendorf™ 022637223
 View more versions of this product
Catalog No. 36-100-7272
Explore available promotions
Description
Description
For use with Standard and Vacutainer conical and micro test tubes.
Specifications
Specifications
Eppendorf™ special rotor for 5804/5804R/5810/5810R microcentrifuge
1.6 cm
2
4 to 10 mL