missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 1 L
Plug-and-play tangential flow filtration cassette that is made from regenerated cellulose, for ultrafiltration and diafiltration of intermediate sample volumes.
$301.80 - $301.80
Specifications
Filter Type | Ultrafiltration |
---|---|
Format | Cassette |
For Use With (Equipment) | Peristaltic Pump |
Material | Polycarbonate |
Material (Membrane) | Regenerated Cellulose |
Catalog Number | Mfr. No. | MWCO | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Price | Quantity | |||||
10-569-116
|
Sartorius
VFS050H0300SV |
300 kDa |
Pack for $301.80
|
|
|||||
10-569-118
|
Sartorius
VFS050H0100SV |
100 kDa |
Pack for $301.80
|
|
|||||
10-569-120
|
Sartorius
VFS050H0030SV |
30 kDa |
Pack for $301.80
|
|
|||||
10-569-122
|
Sartorius
VFS050H0010SV |
10 kDa |
Pack for $301.80
|
|
|||||
10-569-124
|
Sartorius
VFS050H0005SV |
5 kDa |
Pack for $301.80
|
|
|||||
10-569-126
|
Sartorius
VFS050H0002SV |
2 kDa |
Pack for $301.80
|
|
|||||
Description
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: Regenerated cellulose
- Recommended feed volume: 500 to 1000 mL
Specifications
Ultrafiltration | |
Peristaltic Pump | |
Regenerated Cellulose | |
Non-sterile | |
1 L |
Cassette | |
Polycarbonate | |
0.5 to 1 L | |
TFF cassette |