PYREX™ Regular Cyanide Distillation Apparatus Replacement Parts
Replacement parts for use with Corning PYREX Regular Cyanide Distillation Apparatus
$177.81 - $522.83
Specifications
For Use With (Application) Pyrex™ cyanide distilling apparatus
Material Borosilicate Glass
View More Specs
Products 3
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
05-723-16D
View Documents
PYREX™
3350DTO
Dispersion Tube; Standard taper inner joint: 29/42
Case for $522.83
05-723-16E
View Documents
PYREX™
3350TO
Trap; Standard taper inner joint: 29/42
Case for $228.44
05-723-16A
View Documents
PYREX™
3350ITO
Inlet Tube; Standard taper inner joint: 19/38
Case for $177.81
Description
Description
For Cyanide Distillation Apparatus, see cat. no. 0572316
Compliance
Meets U.S. EPA SW-846 Methods 9010 and 9012 and ASTM™ Method D 2036.
Specifications
Specifications
Pyrex™ cyanide distilling apparatus
Borosilicate Glass
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'