Reichert™ Darkfield Quebec™ Colony Counters
Count bacterial colonies quickly and accurately
Supplier:  Reichert™ 13332500
 View more versions of this product
Catalog No. RE-3325
Explore available promotions
Includes: Both models are supplied with white-ruled Wolffheugel guide plate, three-wire cord and 1.5X lens. Electronic Register model also includes probe and ground.
Description
Description
Exclusive darkfield design provides even, glare-free illumination. Contrasted against darkfield background, colonies are bright—readily distinguished from other structures in agar. Each probing action into the colony completes a circuit and is automatically recorded (Electronic Register model only).
Key Features (both models):
  • Highly polished, annular reflector spreads light from 40-watt bulb uniformly over entire culture plate
  • Operating switch conveniently mounted on front of instrument case
  • Hinged door provides easy access to change light bulb—just remove two screws
  • Container holder adapts for centering round dishes up to 100mm diameter or square culture plates up to 100 × 100mm
  • Adjustable focusing rod allows 1.5X lens to be raised or lowered for magnified viewing
  • Lens rotates 360° for ready access to culture plates
  • Reset bar provides immediate zeroing of five-digit counter
  • Built-in tilt leg may be mounted in front or rear; allows convenient tilt angle or may be locked flat to instrument base
Electronic register model
  • Operated by needle-point probe with magnetic ground
  • When ground and counting probes both touch conductive agar in dish or plate, an electrical circuit is completed through the integral amplifier to activate the digital counter
Certifications
UL Listed
Specifications
Specifications
Darkfield Colony Counter
10 in.
10.5 in.
9.25 in.
12 lb.
60Hz
UL
USA cord, Wolffheugel counting plate, three-wire cord and 1.5X lens
Standard
25cm
27cm
23cm
5500g
115V
Count bacterial colonies quickly and accurately