missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Reichert™ Vet360-Chek Digital Handheld Refractometer
Automatic, multi-scale clinical refractometer for veterinary use
Supplier: Reichert™ 13940018
Includes: 2 AAA batteries
Description
- Developed by using appropriate chemistry data for large and small animals
- Four measurement scales (canine urine specific gravity, feline urine specific gravity, large animal urine specific gravity, serum protein concentration of all animals)
- Can be quickly accessed with simple push of button
- Digital technology gives accurate and precise measurement in seconds
- Automatic temperature compensated measurements
- 5-digit, full view LCD screen
- Compact with detachable neck lanyard, rubber side grips for easy handling
For over 10000 measurements
With auto-off sleep mode
Ratings: IP65 Dustproof/Water Resistant
Certifications
CE marked
Compliance
RoHS, WEEE
Warranty
One year (factory)
Specifications
1 to 1.06 Canine Urine Specific Gravity, 1 to 1.09 Feline Urine Specific Gravity, 1 to 1.061 Large Animal Urine Specific Gravity, 2.5 to 15 Serum Protein | |
0.005 USG canine, 0.005 USG feline, 0.005 USG large animal, 0.1g/100mL Serum Protein | |
2 AAA Batteries | |
3.5 oz. | |
20°C |
Distilled Water | |
589nm LED | |
IP65, RoHS, WEEE, CE | |
100 g |