Learn More
Thermo Scientific™ ALA Disk
Rapidly differentiate Haemophilus species on the basis of the detection of porphyrin synthesis using ALA Disk.
Supplier: Thermo Scientific™ RE21115
Description
Rapidly differentiate Haemophilus species on the basis of the detection of porphyrin synthesis using Thermo Scientific™ Remel™ ALA Disk. In 1974, Kilian described a rapid method for identification of X-factor independent Haemophilus spp. based on the ability to synthesize heme precursors from ALA1. Lund and Blazevic determined the porphyrin test is more rapid and accurate for identification of X-factor-independent strains of Haemophilus (e.g. H. parainfluenzae and H. parahaemolyticus) than the satellite test2.
Members of the genus Haemophilus are small non-motile Gram-negative bacilli traditionally defined by a requirement for X and/or V factor3. X factor is derived from hemoglobin and V factor through the satellite relationship with Staphylococcus aureus.
Rapidly differentiate Haemophilus species on the basis of their detection of porphyrin synthesis using ALA Disk.
- Ready to use: Reagent-impregnated disk
- Rapid test: Results are obtained within a few hours.
- Accurate test: For detection of porphyrins which are only excreted by X-factor-independent Haemophilus spp.
- Easy to identify: A positive result showcases bright yellow fluorescence under long wave ultraviolet light.
After incubation the disk is exposed to long wave ultraviolet light in a dark room for any indication of fluorescence. Biberstein et al. studied the action of Haemophilus on δ-aminolevulinic acid (ALA) and reported a perfect correlation between X-factor-independence and the ability to convert ALA to porphyrins4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
ALA Disk™ | |
Porphyrin | |
ALA Disk | |
25 Disks/Vial |