Learn More
Thermo Scientific™ Remel™ Anaerobic Blood Agar, Monoplate
Isolate and cultivate anaerobic organisms, including fastidious strains with this ready-to-use medium.
$135.05 - $135.05
Description
Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance.
Anaerobe Blood Agar contains peptones which supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K, and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium.
Specifications
| Anaerobic Blood Agar | |
| Agar |