Learn More
Thermo Scientific™ Remel™ BactiCard™ E. coli
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
Supplier: Thermo Scientific™ R21108
Description
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
CPT Code: 87147
CLIA Complexity: Non-waived
- Two separate test circles for MUG and Indole
- Results after 15 minutes of incubation
- Positive MUG test displays fluorescence under longwave ultraviolet light
- Positive Indole test evidenced by a color change to redv
- If indole and MUG are positive, then E. coli is present
Includes:
Test cards, rehydrating fluid, MUG reagent, and applicator sticks.
Specifications
2°C to 8°C | |
E. coli | |
1 Kit | |
15 min. |
BactiCard™ E. coli | |
BactiCard | |
25 Tests/Kit |
For in vitro diagnostic use.