Thermo Scientific™ Remel™ BactiCard™ Neisseria
Uses enzyme technology for presumptive identification of Neisseria spp. and Moraxella catarrhalis on selective media.
Supplier:  Thermo Scientific™ R21110
Catalog No. R21110
Explore available promotions
Description
Description
Thermo Scientific™ Remel BactiCard Neisseria uses enzyme technology for presumptive identification of pathogenic Neisseria spp. and Moraxella catarrhalis isolated on selective media.
Specifications
Specifications
BactiCard™ Neisseria
Neisseria species , M. catarrhalis
Enzyme Immunoassay Detection
Pathogenic Neisseria Spp., Moraxella Catarrhalis
Yes
Bacticard Neisseria
25 Tests/Kit