Thermo Scientific™ Burkholderia Cepacia Selective Agar
Selectively isolate and differentiate Burkholderia cepacia complex from respiratory secretions of patients with cystic fibrosis with this medium.
Supplier:  Thermo Scientific™ RE01245
Catalog No. R01245
Explore available promotions
Description
Description
Selectively isolate and differentiate Burkholderia cepacia complex from respiratory secretions of patients with cystic fibrosis with Thermo Scientific™ Remel™ Burkholderia Cepacia Selective Agar prepared plates. Selective agents crystal violet, polymixin B, gentamicin, and vancomycin inhibit organisms commonly found in respiratory secretions other than B. cepacia complex. Sucrose and lactose are carbohydrates for enrichment and differentiation with phenol red as a pH indicator. B. cepacia appear as yellow colonies on the medium.
Since the mid 1980’s, B.cepacia complex has been recognized as an important pathogen affecting patients with cystic fibrosis. Colonization and pulmonary infection with this organism have been associated with a rapid decline in pulmonary function and poor patient prognosis. 
Specifications
Specifications
Burkholderia Cepacia Selective Agar (BCSA)
12mm x 85mm Monoplate
Agar
10/Pk.