Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Supplier:  Thermo Scientific™ RE112520
 View more versions of this product
Catalog No. R112520
Explore available promotions
Description
Description
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Not intended for IVD use.
Specifications
Specifications
Industrial Reference: BAM, USDA, USP.
PB -1000mL
Each
R112520
Phosphate Buffer, Butterfield's
Buffer Solution
1000 mL