Thermo Scientific™ Remel™ Calibrated Inoculating Loops
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Supplier:  Thermo Scientific™ RE501510
 View more versions of this product
Catalog No. R501510
Explore available promotions
Description
Description
Thermo Scientific™ Remel Calibrated Inoculating Loops are available in platinum, twisted nichrome and and plastic. Some are color coded with insulated handle.
  • Twisted nichrome
  • With or without insulated handle
  • Color-coded
Specifications
Specifications
Plastic
Yellow
960/Cs.
10 μL
Inoculating Loop